ContactUsLinkComponent
Escherichia coli WDCM 00090 Vitroids 50,000-150,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000907-10EA
Supplier:  Merck
New Product
Escherichia coli WDCM 00090 Vitroids 50,000-150,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000907-10EA
Supplier:  Merck