ContactUsLinkComponent
ActivArmr 47401 - Abrasion-resistant gloves offering an ideal alternative to PVC and light leather gloves - 100% cotton interlock liner: Superior abrasion resistance - ¾ dipped nitrile coating: Outstanding oil-repellent gloves - Elasticated knitwris 1 * 1 PAIR
Catalog # ANSE47-401/11
Supplier:  Ansell
New Product
ActivArmr 47401 - Abrasion-resistant gloves offering an ideal alternative to PVC and light leather gloves - 100% cotton interlock liner: Superior abrasion resistance - ¾ dipped nitrile coating: Outstanding oil-repellent gloves - Elasticated knitwris 1 * 1 PAIR
Catalog # ANSE47-401/11
Supplier:  Ansell