ContactUsLinkComponent
Muffle Furnace LT15/11/B510 Tmax1100 with lift door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
Catalog # 455-2839
Supplier:  NABERTHERM
New Product
Muffle Furnace LT15/11/B510 Tmax1100 with lift door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
Catalog # 455-2839
Supplier:  NABERTHERM