Muffle Furnace LT9/12/B510 Tmax1200 with lift door and protective gas connection and over-temperature limiter 1 * 1 items
455-2789
New Product
Muffle Furnace LT9/12/B510 Tmax1200 with lift door and protective gas connection and over-temperature limiter 1 * 1 items
455-2789
New Product
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection