SteadySpray Electrode High Micro 50-200µL/min 2-pack of electrodes for use with the OptiFlow Turbo V source. To be used with flow rates 50-200 uL/min. Stainless steel, tan sleeve. 1 * 2 items
Catalog # EKSI5061576
Supplier: EKSIGENT