ContactUsLinkComponent
Human Glucose Transporter GLUT3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Glucose Transporter GLUT3 in serum, plasma,tissue homogenates, cell lysates or other bio 1 * 48 Tests
Catalog # ANTIA2716-48
Supplier:  Antibodies.com
New Product
Human Glucose Transporter GLUT3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Glucose Transporter GLUT3 in serum, plasma,tissue homogenates, cell lysates or other bio 1 * 48 Tests
Catalog # ANTIA2716-48
Supplier:  Antibodies.com