ContactUsLinkComponent
Rat S100 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat S100 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA270671-48
Supplier:  Antibodies.com
New Product
Rat S100 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat S100 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA270671-48
Supplier:  Antibodies.com