ContactUsLinkComponent
Human Enkephalin/ENK ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of human Enkephalin/ENK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or ot 1 * 48 Tests
Catalog # ANTIA2236-48
Supplier:  Antibodies.com
New Product
Human Enkephalin/ENK ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of human Enkephalin/ENK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or ot 1 * 48 Tests
Catalog # ANTIA2236-48
Supplier:  Antibodies.com