ContactUsLinkComponent
Canine IL-18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of canine IL-18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 48 Tests
Catalog # ANTIA2561-48
Supplier:  ANTIBODIES.COM
New Product
Canine IL-18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of canine IL-18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 48 Tests
Catalog # ANTIA2561-48
Supplier:  ANTIBODIES.COM