ContactUsLinkComponent
Rat Uteroglobin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Uteroglobin in serum, plasma, tissue homogenates, cell lysates, lung lavage fluid, cell culture supernates 1 * 48 Tests
Catalog # ANTIA2007-48
Supplier:  Antibodies.com
New Product
Rat Uteroglobin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Uteroglobin in serum, plasma, tissue homogenates, cell lysates, lung lavage fluid, cell culture supernates 1 * 48 Tests
Catalog # ANTIA2007-48
Supplier:  Antibodies.com