ContactUsLinkComponent
Mouse IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant mouse IL-1a ELISA standard, and standard reconstitution buffer. This matched an 1 * 576 Tests
Catalog # ANTIA270519-696
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant mouse IL-1a ELISA standard, and standard reconstitution buffer. This matched an 1 * 576 Tests
Catalog # ANTIA270519-696
Supplier:  ANTIBODIES.COM