ContactUsLinkComponent
Human IL-17F ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-17F in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 48 Tests
Catalog # ANTIA2211-48
Supplier:  Antibodies.com
New Product
Human IL-17F ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-17F in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 48 Tests
Catalog # ANTIA2211-48
Supplier:  Antibodies.com