ContactUsLinkComponent
Rat Adenine Nucleotide Translocator 2/ANT 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in v itro quantitative determination of rat Adenine Nucleotide Translocator 2/ANT 2 in serum, plasma, tissue homogenates, 1 * 96 Tests
Catalog # ANTIA247059-96
Supplier:  ANTIBODIES.COM
New Product
Rat Adenine Nucleotide Translocator 2/ANT 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in v itro quantitative determination of rat Adenine Nucleotide Translocator 2/ANT 2 in serum, plasma, tissue homogenates, 1 * 96 Tests
Catalog # ANTIA247059-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.