ContactUsLinkComponent
Human Carboxypeptidase B2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Carboxypeptidase B2 in plasma. 1 * 96 Tests
Catalog # ANTIA2077-96
Supplier:  ANTIBODIES.COM
New Product
Human Carboxypeptidase B2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Carboxypeptidase B2 in plasma. 1 * 96 Tests
Catalog # ANTIA2077-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.