Human Kallikrein 10/K10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Kallikrein 10/K10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 48 Tests
Catalog # ANTIA2657-48
Supplier: Antibodies.com