ContactUsLinkComponent
Human CD62E ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CD62E in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 96 Tests
  ANTIA1685-96
New Product
Human CD62E ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CD62E in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 96 Tests
  ANTIA1685-96
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.