Human Enteric Cytopathic Human Orphan Virus IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Enteric Cytopathic Human Orphan Virus IgG in serum, plasma, tissue homogenates, and 1 * 96 Tests
Catalog # ANTIA247218-96
Supplier: ANTIBODIES.COM