ContactUsLinkComponent
Human alpha Endorphin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative d etermination of human alpha Endorphin in serum, plasma, tissue homogenates, cell culture supernates or other biologic 1 * 96 Tests
Catalog # ANTIA2639-96
Supplier:  ANTIBODIES.COM
New Product
Human alpha Endorphin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative d etermination of human alpha Endorphin in serum, plasma, tissue homogenates, cell culture supernates or other biologic 1 * 96 Tests
Catalog # ANTIA2639-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.