ContactUsLinkComponent
Human PTGIS/PGIS ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human PTGIS/PGIS in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2590-48
Supplier:  Antibodies.com
New Product
Human PTGIS/PGIS ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human PTGIS/PGIS in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2590-48
Supplier:  Antibodies.com