ContactUsLinkComponent
Full-length soluble protein with foldon trimerization motif, mutated Furin recognition site and 4 stabilising mutations (A892P, A942 P, K986P and V987P), based on/modified from Amanat et al, 2020 (PMID: 32398876) and Hsieh et al, 2020 (PMID: 32703906 1 * 100 µG
Catalog # ANTIA269990-100
Supplier:  ANTIBODIES.COM
New Product
Full-length soluble protein with foldon trimerization motif, mutated Furin recognition site and 4 stabilising mutations (A892P, A942 P, K986P and V987P), based on/modified from Amanat et al, 2020 (PMID: 32398876) and Hsieh et al, 2020 (PMID: 32703906 1 * 100 µG
Catalog # ANTIA269990-100
Supplier:  ANTIBODIES.COM