ContactUsLinkComponent
Mouse Viperin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse Viperin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA271039-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Viperin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse Viperin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA271039-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.