ContactUsLinkComponent
Bovine alpha 1 Acid Glycoprotein/AGP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of bovine alpha 1 Acid Glycoprotein/AGP in serum, plasma, urine or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1971-48
Supplier:  Antibodies.com
New Product
Bovine alpha 1 Acid Glycoprotein/AGP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of bovine alpha 1 Acid Glycoprotein/AGP in serum, plasma, urine or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1971-48
Supplier:  Antibodies.com