ContactUsLinkComponent
Human AACT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human AACT in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2144-96
Supplier:  ANTIBODIES.COM
New Product
Human AACT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human AACT in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2144-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.