ContactUsLinkComponent
Rat ZnT3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat ZnT3 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA270994-96
Supplier:  ANTIBODIES.COM
New Product
Rat ZnT3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat ZnT3 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA270994-96
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.