Human alpha MSH ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of human alpha MSH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolog 1 * 48 Tests
Catalog # ANTIA2392-48
Supplier: Antibodies.com