Human GM-CSF Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-AP, recombinant human GM-CSF ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 576 Tests
Catalog # ANTIA270389-696
Supplier: Antibodies.com