ContactUsLinkComponent
Human Carbonic Anhydrase 1/CA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Carbonic Anhydrase 1/CA1 in serum, plasma, tissue homogenates, cell lysates or other biol 1 * 48 Tests
Catalog # ANTIA1729-48
Supplier:  Antibodies.com
New Product
Human Carbonic Anhydrase 1/CA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Carbonic Anhydrase 1/CA1 in serum, plasma, tissue homogenates, cell lysates or other biol 1 * 48 Tests
Catalog # ANTIA1729-48
Supplier:  Antibodies.com