ContactUsLinkComponent
Mouse pan IFN alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, recombinant mouse IFN alpha ELISA standard, and standard reconstitution buffer. This mat 1 * 576 Tests
Mouse pan IFN alpha Matched Antibody Pair Kit
Catalog # ANTIA270529-696
Supplier:  Antibodies.com
New Product
CAS Number:  
Mouse pan IFN alpha Matched Antibody Pair Kit
Catalog # ANTIA270529-696
Supplier:  Antibodies.com
New Product
CAS Number:  
Some Products May Appear Restricted