ContactUsLinkComponent
Rat Nicotinic Acetylcholine Receptor alpha 1/CHRNA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Nicotinic Acetylcholine Receptor alpha 1/CHRNA1 in tissue homogenates o 1 * 48 Tests
Catalog # ANTIA2591-48
Supplier:  ANTIBODIES.COM
New Product
Rat Nicotinic Acetylcholine Receptor alpha 1/CHRNA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Nicotinic Acetylcholine Receptor alpha 1/CHRNA1 in tissue homogenates o 1 * 48 Tests
Catalog # ANTIA2591-48
Supplier:  ANTIBODIES.COM