ContactUsLinkComponent
Rat nNOS (neuronal) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of rat nNOS (neuronal) in tissue homogenates, cell lysates, cell culture supernates or other biological fluid 1 * 48 Tests
Catalog # ANTIA1967-48
Supplier:  Antibodies.com
New Product
Rat nNOS (neuronal) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of rat nNOS (neuronal) in tissue homogenates, cell lysates, cell culture supernates or other biological fluid 1 * 48 Tests
Catalog # ANTIA1967-48
Supplier:  Antibodies.com