ContactUsLinkComponent
Mouse IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant mouse IL-1a ELISA standard, and standard reconstitution buffer. This matched an 1 * 1.920 Tests
Catalog # ANTIA270519-2096
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-1 alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-HRP, recombinant mouse IL-1a ELISA standard, and standard reconstitution buffer. This matched an 1 * 1.920 Tests
Catalog # ANTIA270519-2096
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.