ContactUsLinkComponent
Bovine Carbonic Anhydrase 1/CA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of bovine Carbonic Anhydrase 1/CA1 in serum, plasma, tissue homogenates, cell lysates or other bi 1 * 48 Tests
Catalog # ANTIA270956-48
Supplier:  Antibodies.com
New Product
Bovine Carbonic Anhydrase 1/CA1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of bovine Carbonic Anhydrase 1/CA1 in serum, plasma, tissue homogenates, cell lysates or other bi 1 * 48 Tests
Catalog # ANTIA270956-48
Supplier:  Antibodies.com