Human Mannan Binding Lectin/MBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Mannan Binding Lectin/MBL in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 48 Tests
Catalog # ANTIA2504-48
Supplier: ANTIBODIES.COM