Chicken IL-18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of chicken IL-18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Catalog # ANTIA2650-48
Supplier: Antibodies.com