Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human alpha 2C Adrenergic Receptor/ADRA2C protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA325091-100
Supplier: ANTIBODIES.COM