ContactUsLinkComponent
Bovine Testosterone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative det ermination of bovine Testosterone in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid 1 * 96 Tests
Catalog # ANTIA326908-96
Supplier:  ANTIBODIES.COM
New Product
Bovine Testosterone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative det ermination of bovine Testosterone in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid 1 * 96 Tests
Catalog # ANTIA326908-96
Supplier:  ANTIBODIES.COM