ContactUsLinkComponent
A heterodimer of IL-23A (Arg20-Pro189) with a C-terminal human Fc tag and IL23B (Ile23-Ser328) with a C-terminal 6xHis tag). The hum an Fc tag consists of amino acids Glu99-Ala330. 1 * 50 µG
Catalog # ANTIA317738-50
Supplier:  ANTIBODIES.COM
New Product
A heterodimer of IL-23A (Arg20-Pro189) with a C-terminal human Fc tag and IL23B (Ile23-Ser328) with a C-terminal 6xHis tag). The hum an Fc tag consists of amino acids Glu99-Ala330. 1 * 50 µG
Catalog # ANTIA317738-50
Supplier:  ANTIBODIES.COM