This Hydroxyl Free Radical Scavenging Capacity Assay Kit is a quantitative assay kit for measuring Hydroxyl Free Radical Scavenging Capacity in animal and plant tissues, serum, plasma, cells, bacteria, cell supernatant, juice, and honey. 1 * 96 Tests
Catalog # ANTIA319693-96
Supplier: ANTIBODIES.COM