ContactUsLinkComponent
A heterodimer of Integrin alpha 2 (Tyr30-Thr1132) with a C-terminal 6xHis tag and Integrin beta 1 (Gln21-Asp728) with a C-terminal h uman Fc tag. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 50 µG
Catalog # ANTIA318046-50
Supplier:  ANTIBODIES.COM
New Product
A heterodimer of Integrin alpha 2 (Tyr30-Thr1132) with a C-terminal 6xHis tag and Integrin beta 1 (Gln21-Asp728) with a C-terminal h uman Fc tag. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 50 µG
Catalog # ANTIA318046-50
Supplier:  ANTIBODIES.COM