ContactUsLinkComponent
Rabbit N-Acetyl beta-D-Glucosaminidase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit N-Acetyl beta-D-Glucosaminidase in serum, plasma, cell culture supernatant, cell 1 * 96 Tests
Catalog # ANTIA326756-96
Supplier:  ANTIBODIES.COM
New Product
Rabbit N-Acetyl beta-D-Glucosaminidase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit N-Acetyl beta-D-Glucosaminidase in serum, plasma, cell culture supernatant, cell 1 * 96 Tests
Catalog # ANTIA326756-96
Supplier:  ANTIBODIES.COM