ContactUsLinkComponent
Human Anti-Acetylated Ornithin Antibodies ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quanti tative determination of human Anti-Acetylated Ornithin Antibodies in serum, plasma, tissue homogenates, and other bio 1 * 96 Tests
Catalog # ANTIA303161-96
Supplier:  Antibodies.com
New Product
Human Anti-Acetylated Ornithin Antibodies ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quanti tative determination of human Anti-Acetylated Ornithin Antibodies in serum, plasma, tissue homogenates, and other bio 1 * 96 Tests
Catalog # ANTIA303161-96
Supplier:  Antibodies.com