Mouse IFNGR1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IFNGR1 in serum, plasma, and other biological fluids. This ELISA kit only requires 25µ l of sample vol 1 * 96 Tests
Catalog # ANTIA323190-96
Supplier: ANTIBODIES.COM