ContactUsLinkComponent
Rabbit monoclonal antibody to JNK1 (phospho Thr183 + Tyr185) + JNK2 (phospho Thr183 + Tyr185) + JNK3 (phospho Thr221 + Tyr223). 1 * 100 µl
Catalog # ANTIA307546-100
Supplier:  ANTIBODIES.COM
New Product
Rabbit monoclonal antibody to JNK1 (phospho Thr183 + Tyr185) + JNK2 (phospho Thr183 + Tyr185) + JNK3 (phospho Thr221 + Tyr223). 1 * 100 µl
Catalog # ANTIA307546-100
Supplier:  ANTIBODIES.COM