This Ca2+/Mg2+-ATPase Activity Assay Kit is a quantitative assay kit for measuring Ca2+/Mg2+-ATPase activity in serum, plasma, animal and plant tissues, cells, cell supernatant, and bacteria. 1 * 48 Tests
Catalog # ANTIA319665-48
Supplier: ANTIBODIES.COM