Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Neurotensin Receptor 1/NTSR1 protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA317299-100
Supplier: ANTIBODIES.COM