ContactUsLinkComponent
A fusion protein consisting of SARS-CoV-2 (Lys310-Leu560 ) Spike Glycoprotein fused to SARS-CoV-2 Membrane Protein (Met1-Lys15 ) wit h a C-terminal 6xHis tag. 1 * 100 µG
Catalog # ANTIA318236-100
Supplier:  ANTIBODIES.COM
New Product
A fusion protein consisting of SARS-CoV-2 (Lys310-Leu560 ) Spike Glycoprotein fused to SARS-CoV-2 Membrane Protein (Met1-Lys15 ) wit h a C-terminal 6xHis tag. 1 * 100 µG
Catalog # ANTIA318236-100
Supplier:  ANTIBODIES.COM