ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Prostaglandin E Receptor EP2/PTGER2 protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA317331-100
Supplier:  ANTIBODIES.COM
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Prostaglandin E Receptor EP2/PTGER2 protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA317331-100
Supplier:  ANTIBODIES.COM