Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human TSH Receptor/TSH-R protein to be purified and analysed in vitro. 1 * 100 µG
Catalog # ANTIA318455-100
Supplier: ANTIBODIES.COM