ContactUsLinkComponent
Human Anti-Human CD28 Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative dete rmination of human Anti-Human CD28 Antibody in serum, plasma, cell culture supernatant, cell or tissue lysate, and ot 1 * 96 Tests
Catalog # ANTIA326370-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-Human CD28 Antibody ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative dete rmination of human Anti-Human CD28 Antibody in serum, plasma, cell culture supernatant, cell or tissue lysate, and ot 1 * 96 Tests
Catalog # ANTIA326370-96
Supplier:  ANTIBODIES.COM