ContactUsLinkComponent
Mouse Osteoprotegerin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse Osteoprotegerin in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#01 1 * 96 Tests
Catalog # ANTIA323204-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Osteoprotegerin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse Osteoprotegerin in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#01 1 * 96 Tests
Catalog # ANTIA323204-96
Supplier:  ANTIBODIES.COM